Learn More
Sino Biological Human Granzyme H Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10348H08H10
A DNA sequence encoding the human Granzyme H (NP_219491.1) (Met 1-Leu 246) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-4687