Learn More
Sino Biological Human IFNA10 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10349H08H100
A DNA sequence encoding the human IFNA10 (NP_002162.1) (Met1-Asp189) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4692