Learn More
Sino Biological Human Syndecan-2 / SDC2 Protein (His Tag, ECD) DFS Item
Manufacturer:  Sino Biological 10355H08H50
A DNA sequence encoding the human SDC2 (NP_002989.2) (Met1-Glu144) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4711