Learn More
Sino Biological Human IL18BPa Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10357H08H200
A DNA sequence encoding the N-terminal domain (Met 1-Gly 192) of human IL18BPa (NP_001034748.1) was fused with the polyhistidine tag at the C-terminus.
Catalog No. 50-161-4717