Learn More
Sino Biological Human TGFBR2 Protein (His  Tag) DFS Item
Manufacturer:  Sino Biological 10358H08B20
A DNA sequence encoding the human TGFBR2 (NP_003233.4) (Met1-Gln166) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4722