Learn More
Sino Biological Human IL-15 / IL15 / Interleukin 15 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10360H07E5
A DNA sequence encoding the human IL15 (NP_000576.1) (Asn18-Ser162) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4730