Learn More
Sino Biological Human HER4 / ErbB4 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10363H03H50
A DNA sequence encoding the extracellular domain (Met 1-Pro 651) of human ErbB4 precursor (NP_001036064.1) was fused with the C-terminal His-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4736