Learn More
Sino Biological Human MOG Protein (aa 30-149, His Tag) DFS Item
Manufacturer:  Sino Biological 10364H08E20
A DNA sequence encoding the extracellular domain of human MOG (NP_996532.2) (Gly 30-Tyr 149) was expressed, fused with a polyhistidine tag at the C-terminus and an additional Met at the N-terminus.
Catalog No. 50-161-4741