Learn More
Sino Biological Human CD32a / FCGR2A Protein (166 Arg, His Tag) DFS Item
Manufacturer:  Sino Biological 10374H08B50
A DNA sequence encoding the extracellular domain (Met 1-Gly 216) of human CD32a (AAH19931.1) precursor was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4766