Learn More
Sino Biological Human PD1 / PDCD1 / CD279 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10377H05H200
A DNA sequence encoding the human PDCD1 (NP_005009.2) (Met1-Gln167) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-4788