Learn More
Sino Biological Human Thrombopoietin / THPO / TPO Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10381H07H20
A DNA sequence encoding the mature form of human THPO (NP_000451.1) (Ser 22-Gly 353) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4797