Learn More
Sino Biological Human Thioredoxin / TXN / SASP Protein DFS Item
Manufacturer:  Sino Biological 10384HNAE200
A DNA sequence encoding the C-terminal segment of human TXN (P10599) (Met 1-Val 105) was expressed and purified.
Catalog No. 50-161-4810