Learn More
Sino Biological Human TNFSF14 / LIGHT / CD258 Protein (ECD, Fc Tag) DFS Item
Manufacturer:  Sino Biological 10386H01H20
A DNA sequence encoding the human TNFSF14 (AAK26160.1) (Asp74-Val240) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4813