Learn More
Sino Biological Human TNFSF14 / LIGHT / CD258 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10386H15H3100
A DNA sequence encoding the human TNFSF14 (AAK26160.1) (Asp74-Val240) was expressed with the Fc region of rabbit IgG at the N-terminus..
Catalog No. 50-161-4821