Learn More
Sino Biological Human CD16a / FCGR3A Protein (176 Val, His Tag) DFS Item
Manufacturer:  Sino Biological 10389H08H150
A DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human CD16a (AAH17865.1) was expressed, with a C-terminal polyhistidine tag. It is identical to FCGR3A158F/V in the reference.
Catalog No. 50-161-4829