Learn More
Sino Biological Human CD16a / FCGR3A Protein (176 Phe, His & AVI Tag), Biotinylated DFS Item
Manufacturer:  Sino Biological 10389H27HB50
A DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human CD16a (P08637-1) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. It is identical to FCGR3A158F/V in the reference.
Catalog No. 50-161-4838