Learn More
Sino Biological Human CD16a / FCGR3A Protein (176 Val) (His & AVI Tag) DFS Item
Manufacturer:  Sino Biological 10389H27H150
A DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human CD16a (AAH17865.1) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. It is identical to FCGR3A158F/V in the reference.
Catalog No. 50-161-4833