Learn More
Sino Biological Human IL6 / Interleukin-6 Protein DFS Item
Manufacturer:  Sino Biological 10395HNAE100
A DNA sequence encoding the mature form of human IL6 (NP_000591.1) (Val 30-Met 212) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-161-4849