Learn More
Sino Biological Human IL20RA / IL-20RA Protein (ECD, Fc Tag) DFS Item
Manufacturer:  Sino Biological 10397H02H50
A DNA sequence encoding the human IL20RA (NP_055247.3) (Met1-Lys250) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4858