Learn More
Sino Biological Human IL6R / CD126 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10398H08H50
A DNA sequence encoding the extracellular domain of human IL6R (NP_000556.1) corresponding to amino acid (Met 1-Pro 365) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4862