Learn More
Sino Biological Human CD4 / LEU3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10400H08H50
A DNA sequence encoding the human CD4 (NP_000607.1) extracellular domain (Met 1-Trp 390) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4868