Learn More
Sino Biological Human KLK-1 / Kallikrein-1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10407H08H20
A DNA sequence encoding the human KLK1 (NP_002248.1) precursor (Met 1-Ser 262) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4875