Learn More
Sino Biological Human TRAIL R1 / CD261 / TNFRSF10A Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10408H02H200
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4877