Learn More
Sino Biological Human TRAIL R1 / CD261 / TNFRSF10A Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10408H03H200
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4879