Learn More
Sino Biological Human TNFSF10 / TRAIL / APO-2L / CD253 Protein DFS Item
Manufacturer:  Sino Biological 10409HNAE10
A DNA sequence encoding the human TNFSF10 (NP_003801.1) (Val 114-Gly 281) with an initial Met was expressed and purified.
Catalog No. 50-161-4882