Learn More
Sino Biological Human KLK7 / PRSS6 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10416H08H20
A DNA sequence encoding the human KLK7 (NP_005037.1) extracellular domain (Met 1-Arg 253) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4896