Learn More
Sino Biological Human TNFR2 / CD120b / TNFRSF1B Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10417H08H50
A DNA sequence encoding the extracellular domain (Met 1-Asp 257) of human TNFRII (NP_001057.1) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-4902