Learn More
Sino Biological Human TNFR2 / CD120b / TNFRSF1B Protein (aa 1-268, 196 Met/Arg, His Tag) DFS Item
Manufacturer:  Sino Biological 10417H08H150
A DNA sequence encoding the human TNFRSF1B (P20333-2) (Met1-Arg268, natural variant 196 Met/Arg) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4901