Learn More
Sino Biological Human MKK6 / MEK6 / MAP2K6 Protein (207 Ser/Asp, 211 Thr/Asp, His & GST Tag) DFS Item
Manufacturer:  Sino Biological 10422H20B120
A DNA sequence encoding the human MAP2K6 isoform 1 (P52564-1) (Met 1-Asp 334, Ser 207/Asp, Thr 211/Asp) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4903