Learn More
Sino Biological Human MKK6 / MEK6 / MAP2K6 Protein DFS Item
Manufacturer:  Sino Biological 10422HNCB20
A DNA sequence encoding the human MKK6 (NP_002749.2) (Met 1-Asp334) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-4909