Learn More
Sino Biological Human MSR1 / SCARA1 / CD204 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10427H07H50
A DNA sequence encoding the extracellular domain (Lys 77-Leu 451) of human scavenger receptor 1 isoform type 1 (NP_619729.1) was fused with the polyhistidine tag at the N-terminus.
Catalog No. 50-161-4921