Learn More
Sino Biological Human / Mouse / Rat / Cynomolgus / Rhesus Activin A / INHBA Protein DFS Item
Manufacturer:  Sino Biological 10429HNAH10
A DNA sequence encoding mature form of human INHBA (NP_002183.1) (Gly311-Ser426) was expressed. The mature form sequences of the human, mouse, rat, cynomolgus and rhesus INHBA are identical.
Catalog No. 50-161-4926