Learn More
Sino Biological Human IGFBP3 / IBP3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10430H07H20
A DNA sequence encoding the mature form of human IGFBP3 (NP_000589.2) (Gly28-Lys 291) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4930