Learn More
Sino Biological Human ACYP2 / Acylphosphatase-2 Protein (GST Tag) DFS Item
Manufacturer:  Sino Biological 10437H09E20
A DNA sequence encoding the human ACYP2 (P14621) (Ser 2-Tyr 99) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-4942