Learn More
Sino Biological Human Biglycan / BGN Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10447H08H50
A DNA sequence encoding the human BGN (NP_001702.1) (Asp38-Lys368) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4962