Learn More
Sino Biological Human Oncostatin M / OSM Protein DFS Item
Manufacturer:  Sino Biological 10452HNAH5
A DNA sequence encoding the human OSM (NP_065391.1) (Met1-Arg221) was expressed.
Catalog No. 50-161-4979