Learn More
Sino Biological Human Contactin 2 / CNTN2 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10457H08H50
A DNA sequence encoding the human CNTN2 (NP_005067.1) precursor (Met 1-Asn 1012) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4983