Learn More
Sino Biological Human Carbonic Anhydrase XIV / CA14 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10458H08H20
A DNA sequence encoding the extracellular domain of human CA14 (NP_036245.1) (Met 1-Met 290) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4985