Learn More
Sino Biological Human HGF / Hepatocyte Growth Factor Protein DFS Item
Manufacturer:  Sino Biological 10463HNAS5
A DNA sequence encoding the human HGF (NP_000592.3) precursor (Met 1-Ser 728) was expressed and purified.
Catalog No. 50-161-5000