Learn More
Sino Biological Human TRAIL R2 / CD262 / TNFRSF10B Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10465H03H200
A DNA sequence encoding the human TNFRSF10B (NP_003833.3) extracellular domain (Met 1-Glu 182) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5006