Learn More
Sino Biological Human Carbonic Anhydrase VIII / CA8 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10469H08E50
A DNA sequence encoding the human CA8 (NP_004047.3) (Met 1-Gln 290) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5019