Learn More
Sino Biological Human Carbonic Anhydrase IV / CA4 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10472H08H20
A DNA sequence encoding the human CA4 (NP_000708.1) (Met1-Lys283) without the pro peptide was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5021