Learn More
Sino Biological Human CCL11 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10474H07E20
A DNA sequence encoding the human CCL11 (P51671) (Gly 24-Pro 97) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5028