Learn More
Sino Biological Human NCR3 / NKp300 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10480H03H20
A DNA sequence encoding the extracellular domain (Met 1-Gly 135) of human NCR3 (NP_667341.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5043