Learn More
Sino Biological Human TNFRSF4 / OX40 / CD134 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10481H03H100
A DNA sequence encoding the extracellular domain (Met 1-Ala 216) of human TNFRSF4 (NP_003318.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5047