Learn More
Sino Biological Human Cathepsin A / CTSA Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10482H08H20
A DNA sequence encoding the human cathepsin A isoform b (Met 1-Tyr 480) (NP_001121167.1) was expressed with a N-terminal signal peptide and a C-terminal polyhistidine tag.
Catalog No. 50-161-5053