Learn More
Sino Biological Human Cathepsin C / CTSC / DPPI Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10484H08H10
A DNA sequence encoding the pro form of human cathepsin C (NP_001805.3) (Met 1-Leu 463) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5056