Learn More
Sino Biological Human Cathepsin S / CTSS Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10487H08H10
A DNA sequence encoding the pro form of human CTSS (NP_004070.3) (Met 1-Ile 331) with a carboxy-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5062