Learn More
Sino Biological Human IL27Ra / TCCR / WSX1 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10489H02H50
A DNA sequence encoding the extracellular domain (Met 1-Arg 514) of human IL27Rα (NP_004834.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5065