Learn More
Sino Biological Human Carboxypeptidase A2 / CPA2 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10499H08H20
A DNA sequence encoding the pre pro form of human CPA2 (NP_001859.1) (Met 1-Tyr 417) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5075